CrossLab Autosampler Syringes and Other Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
4710003200 1510146800 210131490 4710002300 5410046300 6610012100 9910116800
CAAG4710003200EA 589.33 CAD
CAAG4710003200 CAAG1510146800 CAAG210131490 CAAG4710002300 CAAG5410046300 CAAG6610012100 CAAG9910116800
CrossLab Autosampler Syringes and Other Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Dispensing syringe and replacement plungersfor use with PSD 100/120 Programmable Sample Dispenser.
  • Solutions are drawn into and held in the dispensing capillary by the action of the syringe
  • All solutions are separated by an air gap in the dispensing capillary to prevent precipitation or other chemical reactions during sampling
  • The collected solutions are then dispensed into the graphite tube located in the furnace workhead
  • PTFE tipped plunger provides good sealing, reducing bubble formation and preventing cross contamination
  • 100 µl syringe provides
More Product Information