CrossLab Autosampler Sampling Capillaries, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
9910115100 9910032300 9910034200
CAAG9910115100PK 501.12 CAD
CAAG9910115100 CAAG9910032300 CAAG9910034200
CrossLab Autosampler Sampling Capillaries, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Dispensing capillary assemblies forAgilent PSD 100/120 Programmable Sample Dispenser.
  • Works well with solvent and acid-resistant sampling components
  • PTFE capillary tips and capillaries reduce contamination
  • Easy replacement by the user
Agilent provides a range of supplies for the PSD 100/120 Programmable Sample Dspenser used with Agilent GFAAS systems, including sample vials, beakers, adapters, dispensing capillary assemblies, dispensing syringe and replacement plungers.​
Dispensing capillary assemblies are made from PTFE tubing, making them idealMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable