CrossLab SIPS 10/20 Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
9910076100 9910077600 6610014600 9910080300
CAAG9910076100KT 106.92 CAD
CAAG9910076100 CAAG9910077600 CAAG6610014600 CAAG9910080300
CrossLab SIPS 10/20 Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Typical supplies for use of SIPS 10/20 sample introduction and dilution system with flame AA systems.
  • The SIPS peristaltic pump tubing specially selected to provide extended lifetime and accurate dilution
  • Unique pump bands are positioned between the presser arm and the peristaltic pump tubing
  • The pump band reduces stretching, improving lifetime of the tubing, and reducing flow pulsation to ensure accurate dilution
  • SIPS Mariotte bottles are constant pressure head bottles, which ensures that the flow of liquid out of the bottle remains constant for, accurate dilution
  • Suitable for storage of diluent, or bulk ionization suppressant solution
  • Barb connectors on the lid allow easy connection to the tee piece
  • The tee piece allows mixing of the sample flow with the diluent
  • The three-way tee used with SIPS 20 allows diluent and modifier/internal standard to be mixed before introduction into the nebulizer
The Sample Introduction Pump System (SIPS) is an innovative sample introduction and dilution system for flame AA. SIPS eliminates the task of manual preparation of multiple calibration standards, providing fast, accurate online dilution of over-range samples.