CrossLab Column Nuts for GC
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0100-1365 0100-1375 0100-1378 19251-60575 19256-60590 5180-4109 5180-4115 5181-1292 19243-80510 8710-0510 05988-20066 5181-8830 05921-21170 18740-20800 5020-8292 5020-8293 5020-8294 5190-1437 5190-1438 G3510-20018 5183-4732 5188-5312 5180-4113 5190-1439 3150-0417
CA0100-1365EA 38.01 CAD
CA0100-1365 CA0100-1375 CA0100-1378 CA19251-60575 CA19256-60590 CA5180-4109 CA5180-4115 CA5181-1292 CA19243-80510 CA8710-0510 CA76508-016 CA76508-102 CA76514-516 CA76514-566 CA76514-602 CA76514-604 CA76514-606 CAAG5190-1437 CAAG5190-1438 CAAGG351020018 CAAG5183-4732 CAAG5188-5312 AG5180-4113 AG5190-1439 AG3150-0417
CrossLab Column Nuts for GC
Fittings and Connectors
Column nuts come in ¹/₁₆" stainless steel, ¹/₈" and ¹/₄" brass, and new self tightening nuts.
  • No retightening means less wasted time
  • Finger-tight design lets anyone make consistent quality connections without tools
  • Low-torque seal prevents ferrules from sticking or crumbling
  • Lower column bleed prolongs column life
One of the most basic tasks in GC, installing the column, can be challenging. The GC oven is a dark, small space, and column installation uses tiny parts that need to be assembled with precision. Installation of the column into both the inlet and detector must be done properly to get accurate and reproducible results. To facilitate good column installation, Agilent offers a selection of capillary column nuts for GC fittings. The latest innovation, the Self Tightening column nut, prevents leaking and the need to retighten when using graphite/polyimide blend ferrules, which are recommended for use with oxygen sensitive detectors such as ECD or mass spectrometry.