CrossLab Column Ferrules for Third Party GC Instruments
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8004-0212 8004-0214 8004-0216 8010-0269 8010-0304 8001-0212 8004-0211 8004-0217 8010-0267 8010-0268 8010-0301 8010-0302 8010-0314 8001-0211 8001-0213 8001-0221 8001-0222 8001-0223 8002-0212 8002-0213 8002-0214 8002-0215 8002-0216 8002-0217 8002-0220 8002-0222 8003-0212 8003-0216 8003-0219 8003-0223 8004-0213 8004-0215 8004-0219 8010-0303 8010-0305 8010-0306 8010-0307 8010-0308 8010-0309 8010-0310 8010-0311 8010-0312 8010-0313 8010-0315 8002-0221
CA8004-0212PK 92.99 CAD
CA8004-0212 CA8004-0214 CA8004-0216 CA8010-0269 CA8010-0304 CAAG8001-0212 CAAG8004-0211 CAAG8004-0217 CAAG8010-0267 CAAG8010-0268 CAAG8010-0301 CAAG8010-0302 CAAG8010-0314 CAAG8001-0211 CAAG8001-0213 CAAG8001-0221 CAAG8001-0222 CAAG8001-0223 CAAG8002-0212 CAAG8002-0213 CAAG8002-0214 CAAG8002-0215 CAAG8002-0216 CAAG8002-0217 CAAG8002-0220 CAAG8002-0222 CAAG8003-0212 CAAG8003-0216 CAAG8003-0219 CAAG8003-0223 CAAG8004-0213 CAAG8004-0215 CAAG8004-0219 CAAG8010-0303 CAAG8010-0305 CAAG8010-0306 CAAG8010-0307 CAAG8010-0308 CAAG8010-0309 CAAG8010-0310 CAAG8010-0311 CAAG8010-0312 CAAG8010-0313 CAAG8010-0315 AG8002-0221
CrossLab Column Ferrules for Third Party GC Instruments
Fittings and Connectors
Column ferrules for PerkinElmer, Thermo Scientific, and Varian/Bruker GC Systems.
  • All column ferrules are nonstick and provide a leak-free seal
  • Graphite ferrules provide an easy-to-use and highly stable seal, boast a high temperature limit, and can be removed easily
  • Vespel/graphite ferrules are mechanically robust and maintain effectiveness over a long lifetime
  • Vespel ferrules are mechanically robust, maintain a long lifetime, and can be reused or removed easily
  • To ensure performance and longevity, column ferrules should be replaced at the same time as
More Product Information