CrossLab Adapters and Unions for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0100-0900 0100-2441 5022-2145 0100-2298 0100-1847 5022-2184 5022-2133 5067-4741 5067-6127 5022-2144 5042-8517 5042-8518 5065-9971
CA0100-0900EA 189.05 CAD
CA0100-0900 CA0100-2441 CA5022-2145 CA0100-2298 CA76514-496 CA76514-622 CA5022-2133 CA5067-4741 CA5067-6127 CAAG5022-2144 CAAG5042-8517 CAAG5042-8518 CAAG5065-9971
CrossLab Adapters and Unions for HPLC, Agilent Technologies
Fittings and Connectors
High-quality LC unions and adapters for a range of equipment and applications.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
Agilent manufacture a wide variety of high-quality LC Unions suitable for a range of equipment and applications, including stainless steel ZDV unions (with and without fittings) for standard LC, ZDV unions (PEEK or stainless steel with PEEK inserts) for bio applications, and high-flow unions (no fittings) for prep LC. A range of adapters and connectors are also available, including barbed Y-connectors, T-connectors, and female-to-female or male-to-female adapters.