CrossLab Reciprocating Dissolution Tubes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
27-1020 27-1000 27-1010 27-1030 G7972-47000
CAAG27-1020EA 149.65 CAD
CAAG27-1020 CA27-1000 CAAG27-1010 CAAG27-1030 CAAGG797247000
CrossLab Reciprocating Dissolution Tubes, Agilent Technologies
Dissolution Apparatus
Several tube vessels are available for Apparatus 3 and 7.
  • Reciprocating Cylinders for BIO-DIS and 400-DS
Ideal for automatic dissolution testing.