CrossLab Reciprocating Dissolution Holders, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
27-0100 27-0102 27-0101 27-0103 27-0104 27-0106 27-0107 27-0108 27-0109 27-3000 27-3001 27-3002 27-8005 27-8020 27-8036 27-8600 27-8620 27-8621 27-8622 33-9007 33-9009 33-9012 33-9018 33-9022 33-9025 33-9029 33-9046 33-9048 33-9051 33-9052
CAAG27-0100EA 686.91 CAD
CAAG27-0100 CAAG27-0102 CAAG27-0101 CAAG27-0103 CAAG27-0104 CAAG27-0105 CAAG27-0106 CAAG27-0107 CAAG27-0108 CAAG27-0109 CAAG27-3000 CAAG27-3001 CAAG27-3002 CAAG27-8005 CAAG27-8020 CAAG27-8036 CAAG27-8600 CAAG27-8620 CAAG27-8621 CAAG27-8622 CAAG33-9007 CAAG33-9009 CAAG33-9012 CAAG33-9018 CAAG33-9022 CAAG33-9025 CAAG33-9029 CAAG33-9046 CAAG33-9048 CAAG33-9051 CAAG33-9052
CrossLab Reciprocating Dissolution Holders, Agilent Technologies
Dissolution Apparatus
Agilent can provide many different dosage form holders for the Apparatus 7.
  • Accessories for the reciprocating holder apparatus 7
  • Simulates the biorelevant conditions for the skin, gastrointestinal tract and tissue required for transdermal, oral drugs and implants with biorelevant temperature agitation rates and retention times
  • Accommodates a variety of dosage form holders including reciprocating disks, angled disks, spring holders, stent holders, and pointed rods
  • Supports typical volume configurations of 5 to 300 ml, depending on instrument
Ideal for automatic dissolution testing of dosage forms that require a change of media, a smaller volume, or more vigorous agitation. The Agilent 400-DS dissolution apparatus 7 enables of small-volume drug-release testing of medical devices such as drug eluting stents (DES).