CrossLab Dissolution Vessels and Carriers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
27-5000 27-5100 27-5120 12-5000V 12-5035V 12-5152V 27-5130 12-5149V 12-5000 12-5010 12-5010V 12-5033 12-5034 12-5035 12-5040 12-5050 12-5055 12-5145 12-5146 12-5147 12-5148 12-5149 12-5153 12-5157 12-5157V 12-5158 12-5158V 12-5159 12-5200 12-5500 12-5505 13-0010 27-6000 27-6050 33-9005
CA27-5000EA 342.14 CAD
CA27-5000 CA27-5100 CA27-5120 CAAG12-5000V CAAG12-5035V CAAG12-5152V CAAG27-5130 CA12-5149V CAAG12-5000 CAAG12-5010 CAAG12-5010V CAAG12-5033 CAAG12-5034 CAAG12-5035 CAAG12-5040 CAAG12-5050 CAAG12-5055 CAAG12-5070 CAAG12-5120 CAAG12-5125 CAAG12-5145 CAAG12-5146 CAAG12-5147 CAAG12-5148 CAAG12-5149 CAAG12-5153 CAAG12-5157 CAAG12-5157V CAAG12-5158 CAAG12-5158V CAAG12-5159 CAAG12-5200 CAAG12-5500 CAAG12-5505 CAAG13-0010 CAAG27-6000 CAAG27-6050 CAAG33-9005
CrossLab Dissolution Vessels and Carriers, Agilent Technologies
Dissolution Apparatus
EaseAlign, TruCenter and TruAlign dissolution vessels are available in a variety of sizes.
  • Vessel volume sizes include 100 and 200 ml, and 1 and 2 L sizes
  • Low actinic-red, peak, and verified versions are available for select volumes
  • Verified dissolution vessels and other accessories are available to support conformance guidelines for mechanical qualification
  • Conversion kits are available to support small- and large-volume dissolution testing
Agilent provides a variety of dissolution vessels like TruAlign, TruCenter, or EaseAlign vessels. These areMore Product Information