CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
17-4001 17-4003 17-4000 17-4005 17-4010 17-4011 17-4020 17-4021 17-4040 17-4045 17-4100 17-4140 17-4004 17-4015 17-4025 17-4050 17-4055 17-4110 17-4120 17-4130 K1200-00855
CA17-4001PK 155.3 CAD
CA17-4001 CA17-4003 CA17-4000 CAAG17-4005 CAAG17-4010 CAAG17-4011 CAAG17-4020 CAAG17-4021 CAAG17-4040 CAAG17-4045 CAAG17-4100 CAAG17-4140 CAAG17-4004 CAAG17-4015 CAAG17-4025 CAAG17-4050 CAAG17-4055 CAAG17-4110 CAAG17-4120 CAAG17-4130 CAAGK120000855
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Dissolution Apparatus
Made of either color-coded UHMWPE or PVDF, the full flow filter prevents clogging.
  • Available in ultrahigh molecular weight polyethylene (UHMWPE) or polyvinylidene fluoride (PVDF)
  • Full flow filter offers increased surface area to prevent clogging
  • Low protein-binding PVDF filter material available
  • Usable with ¹/₈" diameter cannulas
  • Available in packs of 100 or 1000
  • Full flow filters are color coded for easy identification
  • Certificates of analysis can be provided on request
  • Standard conventional filter tips are made of polyethylene fluoride and can also be
More Product Information