Mega Bond Elut Flash Cartridges, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12256026 12256020 12256042 12256068 12256070 12256074 12256080
CAAG12256026PK 257.96 CAD
CAAG12256026 CAAG12256020 CAAG12256042 CAAG12256068 CAAG12256070 CAAG12256074 CAAG12256080
Mega Bond Elut Flash Cartridges, Agilent Technologies
Chromatography Columns
Mega Bond Elut Flash cartridges offer excellent performance when purifying organic compounds.
  • Convenient, disposable cartridges eliminate the need for packing glass columns
  • Flexible ‘open’ tube design for either liquid or solid samples
  • Reliable, consistent flow characteristics deliver high-resolution performance
Mega Bond Elut Flash cartridges offer excellent levels of performance and productivity for the purification of organic compounds, but also for scaled-up SPE. Prepacked, disposable cartridges offer greater convenience than glass columns that must be washed, dried, and repacked after every sample.