Bond Elut SI, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12102037 12113010 12166008B 12256008 14256008 14102010 14102037
CAAG12102037PK 233.98 CAD
CAAG12102037 CAAG12113010 CAAG12166008B CAAG12256008 CAAG14256008 CAAG14102010 CAAG14102037
Bond Elut SI, Agilent Technologies
Chromatography Columns
Bond Elut SI can be used to separate compounds with highly similar structures.
  • Highly polar phase retains polar molecules from nonpolar matrices
  • High-purity silica
  • Separate compounds with very similar structures
Bond Elut SI is a very polar sorbent and is particularly effective at separating compounds with a very similar structure. Applying the analytes in a nonpolar solvent, then increasing the solvent polarity by increasing the concentration of a polar modifier, such as THF or ethyl acetate, delivers effective separations.