CrossLab Bond Elut SCX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12102013 12102040 12162040B 12256053 12256054 14102040 14102075 14113039 14256011 14256019 14256027 14256035
CAAG12102013PK 388.94 CAD
CAAG12102013 CAAG12102040 CAAG12162040B CAAG12256053 CAAG12256054 CAAG14102040 CAAG14102075 CAAG14113039 CAAG14256011 CAAG14256019 CAAG14256027 CAAG14256035
CrossLab Bond Elut SCX, Agilent Technologies
Chromatography Columns
Bond Elut SCX is a strong cation exchanger with a very low pKa and aromatic functionality.
  • Useful for compounds with cationic and nonpolar characteristics
  • Superior cleanup from a single sorbent
  • Very low pKa creates strong analyte interaction
Bond Elut SCX is a strong cation exchanger with a very low pKa. Although the pKa is similar to Bond Elut PRS, the presence of the benzene ring in the functional group increases the potential for nonpolar interactions. This nonpolar characteristic becomes important in ion exchange from aqueous systems.
Order Now
SPECIFICATIONS
Environmentally Preferable