Bond Elut PPL, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12105003 12256087
CAAG12105003PK 477.93 CAD
CAAG12105003 CAAG12256087
Bond Elut PPL, Agilent Technologies
Chromatography Columns
Bond Elut PPL is a modified styrene-divinylbenzene polymer that retains polar analytes.
  • Modified styrene-divinylbenzene polymer
  • Large particle size enables fast extraction speeds
  • High surface area and capacity for polar analytes
Bond Elut PPL (Priority PolLutant) is a styrene-divinylbenzene (SDVB) polymer modified with a proprietary nonpolar surface. PPL will retain even the most polar classes of analytes. The large particle size eases flow for viscous or particulate-rich water samples, while the large surface area and strong hydrophobicity ensure reproducible extractions with high recoveries.