Bond Elut Plexa PAX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12107206 12257506
CAAG12107206PK 318.99 CAD
CAAG12107206 CAAG12257506
Bond Elut Plexa PAX, Agilent Technologies
Chromatography Columns
Bond Elut Plexa PAX nonpolar/anion-exchange SPE is suitable for polar and nonpolar acidic analytes.
  • Mixed-mode nonpolar polymeric anion exchanger offers high level of analyte selectivity
  • Exclusion of endogenous interferences offers superior cleanliness and minimizes ion suppression
  • Simple, single method for ease-of-use reduces method development time
Bond Elut Plexa PAX lifts the performance standard in analyte cleanup and reproducibility for polar and nonpolar acidic analytes. Plexa PAX particles are functionalized using a proprietary process,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable