Bond Elut Plexa, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12259506 12109610 327832
CA12259506PK 473.99 CAD
CA12259506 CAAG12109610 CAAG327832
Bond Elut Plexa, Agilent Technologies
Chromatography Columns
Bond Elut Plexa offers simple extraction, minimal method development, and improved cleanliness.
  • Nonpolar retention mechanism
  • Improved extract cleanliness minimizes sample matrix interferences
  • Simple methods are amenable to a very broad range of analytes
  • Fast flow, reproducible performance, and ease-of-use
Bond Elut Plexa offers easy-to-use methods for general purpose extraction. A unique polymeric architecture with a nonretentive, hydroxylated, amide-free surface, and a nonpolar PS-DVB core retains small molecules. Binding of proteins and lipids onMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Polarity Polar stationary phase