Bond Elut PH, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12102032 12102062 14102032 14102062 14113005 14256004
CAAG12102032PK 242.98 CAD
CAAG12102032 CAAG12102062 CAAG14102032 CAAG14102062 CAAG14113005 CAAG14256004
Bond Elut PH, Agilent Technologies
Chromatography Columns
Bond Elut PH is a moderately nonpolar sorbent commonly used for nonpolar extractions.
  • Added selectivity compared to other nonpolar sorbents
  • Enhanced retention of planar, conjugated organic molecules
  • Similar polarity to C8
Bond Elut PH is a nonpolar bonded silica material that exhibits a selectivity different from alkyl or aliphatic-functionalized phases, such as C8 or cyclohexyl. The electron density in the aromatic ring enhances retention of conjugated or aromatic ring-containing analytes due to desirable pi-pi interactions.