CrossLab Bond Elut Focus, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
A5106010 A5306022
CAAGA5106010PK 565.76 CAD
CAAGA5106010 CAAGA5306022
CrossLab Bond Elut Focus, Agilent Technologies
Chromatography Columns
Bond Elut Focus delivers outstanding retention for polar and nonpolar analytes.
  • Simple, universal method reduces method validation times
  • 'Power rinse' delivers cleaner extracts, improving sensitivity
  • Hydrogen bond-donor capability increases retention of basic analytes
Bond Elut Focus features a unique amide-functionalized, polar-enhanced sorbent technology delivering outstanding retention. The extraction method eliminates the complex pH adjustments necessary with ion exchange and mixed mode methods. The unique ligand chemistry on the sorbent dramatically increases the retention of basic analytes.
The improved retention for polar analytes with Focus allows the use of a strong 'power rinse' without causing analyte loss. Rigorous washes using 10 to 20% MeOH can be used to aggressively remove endogenous matrix interferences. For very polar analytes that are held by strictly polar interactions, washes of up to 100% can be used.