Florisil, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12113049 12256022 12256030 12256038 12256046 52256014
CAAG12113049PK 223.97 CAD
CAAG12113049 CAAG12256022 CAAG12256030 CAAG12256038 CAAG12256046 CAAG52256014
Florisil, Agilent Technologies
Chromatography Columns
Bond Elut Florisil is particularly ideal for isolating polar compounds from viscous matrices.
  • For cleanup of polar interferences from nonpolar samples
  • Economical
  • Fast flow, ideal for viscous samples
Florisil is a magnesia-loaded silica gel, extremely polar in nature and ideal for the isolation of polar compounds from nonpolar matrices. The larger particle size of the sorbent enables fast flow for large sample volumes. Florisil can therefore be an attractive alternative to silica if the sample matrix is particularly viscous.