Bond Elut Certify II, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12102080 12113051 14113051 14102084 14102088
CAAG12102080PK 234.98 CAD
CAAG12102080 CAAG12113051 CAAG14113051 CAAG14102084 CAAG14102088
Bond Elut Certify II, Agilent Technologies
Chromatography Columns
Bond Elut Certify II is a SAX/nonpolar SPE phase for extracting acidic analytes from biomatrices.
  • Ideal for nonpolar and anionic compounds
  • Optimized for acidic drug analysis
  • Bimodal, nonpolar and strong anion exchange
Bond Elut Certify II was developed specifically for the rapid extraction of acidic drugs and metabolites from urine and other biological matrices. Certify II is a mixed-mode cartridge packed with nonpolar C8 and SAX sorbent. It has been optimized for acidic drugs and other compounds that possess both nonpolar and anionic characteristics.