Bond Elut Certify, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12113054 12256145 12256146 14102051 14113054 52102051 14102085 14113052 52102081 52113051
CAAG12113054PK 249 CAD
CAAG12113054 CAAG12256145 CAAG12256146 CAAG14102051 CAAG14113054 CAAG52102051 CAAG14102085 CAAG14113052 CAAG52102081 CAAG52113051
Bond Elut Certify, Agilent Technologies
Chromatography Columns
Bond Elut Certify is a mixed-mode phase ideal for extraction of basic drugs from urine and blood.
  • Mixed-mode sorbent bed
  • Broad application range for aqueous extraction
  • Bimodal, nonpolar, and strong cation exchange
The Bond Elut Certify cartridge utilizes a packed bed consisting of a nonpolar C8 sorbent and a strong cation-exchange (SCX) sorbent. Certify is most used to extract basic (cationic) drugs from urine and blood. It is also very effective for extracting a wide range of compounds from a diverse number of aqueous matrices.