Bond Elut Alumina, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12102049 12256086 12256043
CAAG12102049PK 283.97 CAD
CAAG12102049 CAAG12256086 CAAG12256043
Bond Elut Alumina, Agilent Technologies
Chromatography Columns
Bond Elut Alumina is a very polar sorbent, with high extraction efficiency even with low bed masses.
  • High extraction efficiency
  • Ideal for electron-rich compounds
  • Better high pH stability than unfunctionalized silica
The small particle size of the Bond Elut Alumina range ensures high extraction efficiency even when small bed masses are used. The alumina surface tends to be slightly more stable under high pH conditions than unfunctionalized silica. Alumina is available in acidic (A), basic (B), and neutral (N) forms.