Bond Elut AccuCAT, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12282001 12282004 12282002 12282005
CAAG12282001PK 364.96 CAD
CAAG12282001 CAAG12282004 CAAG12282002 CAAG12282005
Bond Elut AccuCAT, Agilent Technologies
Chromatography Columns
Bond Elut AccuCAT contains SAX and SCX sorbents to extract acidic, basic, and neutral analytes.
  • SCX and SAX functionalities offer broad analyte extraction potential
  • Ultraclean, mixed sorbent bed delivers for reproducible extractions
  • Compatible with many biological fluids for easy method transfer
Bond Elut AccuCAT cartridges are mixed-mode SPE packed bed cartridges consisting of a strong cation exchange (SCX) and a strong anion exchange (SAX) sorbent. AccuCAT is effective for the extraction of acidic, basic, and neutral analytes from biological samples, particularly for catecholamine extraction from biofluids.