In-line Gas Purifiers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5060-9084 BOT-2 BOT-4 MT120-2 MT200-4 MT400-2 OT3-4 OT3-4-SS 5060-9096 5182-9201 5182-9401 5182-9402 5182-9411 HMT200-4 HT200-2 HT200-4 MT120-4 MT200-2 OT1-2 RMSA-2 RMSH-2-SS RMSH-4-SS 5182-9202 HT3-2 HT3-4 MT-2-SS MT-4-SS OT-2-SS OT-4-SS RMSN-4
CA5060-9084EA 639.74 CAD
CA5060-9084 CABOT-2 CABOT-4 CAMT120-2 CAMT200-4 CAMT400-2 CAOT3-4 CAOT3-4-SS CA5060-9096 CAAG5182-9201 CAAG5182-9401 CAAG5182-9402 CAAG5182-9411 CAAGHMT200-4 CAAGHT200-2 CAAGHT200-4 CAAGMT120-4 CAAGMT200-2 CAAGOT1-2 CAAGRMSA-2 CAAGRMSH-2-SS CAAGRMSH-4-SS CAAG5182-9202 CAAGHT3-2 CAAGHT3-4 CAAGMT-2-SS CAAGMT-4-SS CAAGOT-2-SS CAAGOT-4-SS CARMSN-4
In-line Gas Purifiers, Agilent Technologies
Filters Gas Filters
Inline GC gas purifiers, traps, and filters for removal of oxygen, moisture, and hydrocarbons.
  • Large traps with 250 psig maximum pressure rating and up to 8 liters/min maximum flow
  • Indicating oxygen and moisture purifiers with 100 psig maximum pressure rating
  • Indicating traps change color when adsorption capacity is depleted
  • Moisture or oxygen permeating into gas stream due to a leak is removed by the trap until it expires
Agilent inline gas purifiers (traps or filters) remove common contaminates such as oxygen, moisture, and hydrocarbons from GCMore Product Information