Gas Clean Purification System, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP17972 CP17969 CP17974 CP17971P CP17977 CP17981 CP179880 CP17990 CP738408 CP7995 CP17973
CACP17972EA 391.33 CAD
CACP17972 CAAGCP17969 CAAGCP17974 CAAGCP17971P CAAGCP17977 CAAGCP17981 CAAGCP179880 CAAGCP17990 CAAGCP738408 CAAGCP7995 CACP17973
Gas Clean Purification System, Agilent Technologies
Filters Gas Filters
Gas filter system for reduced risk of column damage, sensitivity loss, and instrument downtime.
  • Delivery of clean gases for accurate analyses
  • Fast, leak-free filter replacement for reduced downtime
  • Economical, with immediate payback
  • Highly sensitive filter indicators provide maximum instrument protection
The Agilent gas clean purification system delivers clean gases, reducing the risks of column damage, sensitivity loss, and instrument downtime. Inserting a gas clean purification system in the gas line immediately before the instrument inlet greatlyMore Product Information