VGA 77 Supplies for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
9910061900 9910062200
CAAG9910061900KT 454.99 CAD
CAAG9910061900 CAAG9910062200
VGA 77 Supplies for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
VGA77 supplies provide the best performance of hydride forming elements such as As, Se, and Hg.
  • Compatible with the VGA 77
  • Tubing kits for VGA conveniently provide all replacement tubing required with the VGA
Vapor generation provides the best performance for analysis of hydride forming elements such as As, Se, and Hg for ICP-OES. VGA 77 supplies such as tubing, gas-liquid seperators, absorption cells, reagents, and maintenance supplies provide the key consumables need to receive optimal performance from your VGA 77. The hydride module for the VGA-77More Product Information