CrossLab Vacuum Pump Supplies for LC/MS, Agilent Technologies
Supplier: Agilent
CrossLab
5063-9153 2710100100 G1960-80039 2710100100 9499342M002 9499342M002 5063-9153 G1960-80039 3162-1057 3162-1057
CAG1960-80039EA 386.4 CAD
AG3162-1057 CAG1960-80039 CAAG3162-1057 CAAG9499342M02 AGG1960-80039 AG5063-9153 CAAG2710100100 AG2710100100 AG9499342M002 CA5063-9153
CrossLab Vacuum Pump Supplies for LC/MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Vacuum pump supplies designed to ensure the accuracy and performance of Agilent's LC/MS systems.
  • Oil mist and filter exhausts for various vacuum pump models
  • Performance maintenance kits for Edwards and MS40+ kits
  • Vacuum and diffusion pump fluid available as well as oil pans and return lines
To ensure the highest levels of accuracy and performance of Agilent's LC/MS systems, a range of vacuum pump supplies is available. These supplies include foreline pump supplies such as oil mist filter kits, cartridge filters, foreline exhaust adapters, and hose clamps.
Foreline and rotary pump oil is also available along with replacement pump oil drip pans. Additional pump supplies and tubing can also be provided. All Agilent vacuum components and replacement parts are manufactured to high levels of quality and consistency.