CrossLab Torch Clamps and Accessories for ICP-OES, Agilent Technologies
Supplier: Agilent
CrossLab
2410023800 2410023800
CAAG2410023800EA 52.83 CAD
CAAG2410023800 AG2410023800
CrossLab Torch Clamps and Accessories for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Torch clamps, bonnets, transfer tubes, cleaning stands, and storage racks for Agilent ICP-OES torches.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
Agilent has a wide variety of ICP-OES torch clamps and torch accessories guaranteed to work with your specific torch and spray chamber setup. Available torch accessories include torch racks, cleaning stands, bonnets and transfer tubes. Also find torch to spray chamber transfer tubes for axial and radial ICP-OES instruments.
The torch cleaning stand for Agilent 5000 series ICP-OES torches, provides a stable platform that allows either the whole torch or the injector/base assembly to be suspended in the cleaning solution. The torch storage rack provides a convient way to store up to five 5000 series ICP-OES torches after cleaning.