CrossLab Spray Chambers for MP-AES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G8000-70007 G8000-70006
CAAGG800070007EA 1606.47 CAD
CAAGG800070007 CAAGG800070006
CrossLab Spray Chambers for MP-AES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Spray chambers for MP-AES instrument.
  • Double pass glass cyclonic spray chamber is the default configuration recommended for most sample types - it provides improved plasma stability with organic and high TDS samples
  • Single pass glass cyclonic spray chamber is recommended when you need improved signal to noise performance with 'clean' matrices
  • Inert spray chamber is recommended for hydrofluoric (HF) acid digests and high TDS samples - an inert torch and nebulizer is also required for HF acid digests
  • Inert spray chamber features a removable baffle providing
More Product Information