CrossLab Spray Chambers for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G8010-60256 2010081700 7910043700 G8010-60271 G8010-68024
CAG8010-60256EA 1943.55 CAD
CAG8010-60256 CAAG2010081700 CAAG7910043700 CAAGG801060271 CAAG9910070400 CAAGG801068024
CrossLab Spray Chambers for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Glass and Inert spray chambers in single and double pass configurations for ICP-OES.
  • Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout
  • Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples
  • Inert spray chamber features a removable baffle providing two spray chambers in one and improves stability with high TDS samples
  • Inert spray chamber is permenantly treated and never need to be sent in for rejuvenation providing more uptime and lower operating costs
  • Inert spray chamber is ideal for Agilent 5100, 5110, 5800, and 5900 ICP-OES and compatible with earlier Agilent 700/Vista and Liberty series instruments with an adapter.
Agilent has a wide range of spray chambers compatible with your Agilent ICP-OES. Glass and inert spray chambers are available in both single and double pass configurations to meet the needs of any sample matrix.
The new Agilent inert spray chamber has a permanently treated sanblasted finish for optimum drainage and continous accurate performance. The inert spray chamber also features a removable baffle allowing both single-pass operation for increased sensitivity and double-pass operation for difficult high-solid matricies.