CrossLab Skimmer Bases for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3280-60608 G3280-60621 G3666-60311 G8400-60625 G1820-65435 G1833-65590 G1833-65591 G3280-60381 G3666-60301 G3666-60401 G3666-60601 G8400-60624 G8400-60626 G8400-60627 G8400-60628 G8400-60634
CAAGG328060608EA 1071.56 CAD
CAAGG328060608 CAAGG328060621 CAAGG366660311 CAAGG840060625 CAAGG182065435 CAAGG183365590 CAAGG183365591 CAAGG328060381 CAAGG366660301 CAAGG366660401 CAAGG366660601 CAAGG840060624 CAAGG840060626 CAAGG840060627 CAAGG840060628 CAAGG840060634
CrossLab Skimmer Bases for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent stainless steel and brass skimmer bases optimize the temperature of the skimmer cone tip.
  • Agilent 7700, 7800, 7900, 8800, 8900 ICP-MS skimmer cone base are avilable in either stainless steel or brass
  • The operating temperature of the skimmer cone tip is controlled by the skimmer base material
  • The stainless steel base is used for nickel (Ni) skimmer cones. The platinum skimmer cone should only be used with the brass skimmer base
  • The ion extraction lenses (x-lens or s-lens) are fixed to the skimmer base
  • The correct type for lens configuration and ICP-MS model (7700, 7800, 7900, 8800, 8900) is required
The ion extraction lenses are fixed to the skimmer base;the correct type for lens configuration and ICP-MS model is required.
The skimmer base ensures the correct operating temperature of the skimmer cone to prevent damage by overheating or increased drift due to excess matrix deposition at the tip if the cone runs too cool. Firmly install the sampling and the skimmer cones on the skimmer base. If the interface cones are not tightly installed on the base, there may be insufficient cooling, which can degrade analytical performance