CrossLab Sampler and Maintenance Kits for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5063-6506 G1329-68727 G4216-68711 G7157-68711
CA5063-6506PK 36.28 CAD
CA5063-6506 CAG1329-68727 CAAGG421668711 CAAGG715768711
CrossLab Sampler and Maintenance Kits for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A comprehensive range of maintenance, accessory, and upgrade kits for HPLC samplers.
  • Noisy baseline, shorter column life, lower analytical sensitivity, and variable retention times can be caused by a lack of preventive maintenance
  • HPLC autosamplers can work at their best with the Agilent preventive maintenance kits
  • Routine sampler maintenance should be done on a regular basis to keep your Agilent LC system performing at its optimum
Agilent provides a comprehensive range of kits for HPLC samplers. Maintenance kits include supplies that require exchanging on a regular basis, accessory kits contain parts to always have on-hand if needed, and upgrade kits enable modification of the original setup.
All parts are manufactured to Agilent's highest levels of quality and have been tested on Agilent instruments to ensure performance that will significantly increase your analytical, instrument, and laboratory efficiency.