CrossLab Sample Trays for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5022-6531 G1364-84516 G1364-84523 G1364-84524 G1364-84525 G7167-60020 G7129-60010 G7129-68210 5023-2471 G1313-27302 G1313-60004 G2258-60111 G4226-60021 G7129-60000 G7129-60110 G9321-60201
CA5022-6531PK 51.92 CAD
CA5022-6531 CAG1364-84516 CAG1364-84523 CAG1364-84524 CAG1364-84525 CAG7167-60020 CAAGG712960010 CAAGG712968210 CAAG5023-2471 CAAGG131327302 CAAGG131360004 CAAGG225860111 CAAGG422660021 CAAGG712960000 CAAGG712960110 CAAGG932160201
CrossLab Sample Trays for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Designed to ensure that your autosampler can accommodate a variety of applications.
  • Agilent provides various trays with different hole sizes and different hole numbers to best suit all your applications
  • Available options include 100 position trays, thermostatted trays, 40 position trays, and 15 position trays for 6 ml vials
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
Agilent autosampler vial trays and drawers are designed to ensure that your autosampler setup isMore Product Information