CrossLab Nebulizers for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G8010-60255 G8010-60293 2010096400 2010106800 G8010-60270 2010126000 CP914506 G8010-60346 G8010-60347
CAG8010-60255EA 2142.82 CAD
CAG8010-60255 CAG8010-60293 CA2010096400 CAAG2010097600 CAAG2010106800 CAAGG801060270 CAAG2010097700 CAAG2010126000 CAAGCP914506 CAAGG801060346 CAAGG801060347
CrossLab Nebulizers for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Glass concentric, Seaspray, Miramist, and inert One Neb 2 nebulizer offering superior performance.
  • Manufactured to fine tolerances for reproducible and stable sample uptake
  • Tight specifications deliver fine droplets for best signal intensity and optimum signal to noise
  • Fewer repeats means less downtime and improved productivity
The universal OneNeb Series 2 nebulizer provides improved sensitivity and presision, greater tolerance to disolved solids, and is inert so it is suitable for use with aggressive acids such as hydrofluoric acid (HF) and mostMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable