CrossLab Manual Injection Valves and Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0101-0376 0101-1242 0101-1243 0101-1244 0101-1245 0101-1246 0101-1248 5067-6845 1400-3166 1535-4046 1535-4048 5063-6502 5065-9922 5067-1503 5067-4191 5067-6831 5067-7031 5067-7032 5067-7033 5068-0321
CA0101-0376EA 103.14 CAD
CA0101-0376 CAAG0101-1242 CAAG0101-1243 CAAG0101-1244 CAAG0101-1245 CAAG0101-1246 CAAG0101-1248 CAAG5067-6845 CAAG1400-3166 CAAG1535-4046 CAAG1535-4048 CAAG5063-6502 CAAG5065-9922 CAAG5067-1503 CAAG5067-4191 CAAG5067-6831 CAAG5067-7031 CAAG5067-7032 CAAG5067-7033 CAAG5068-0321
CrossLab Manual Injection Valves and Supplies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Industry-leading manual injection valves and related high-quality supplies.
  • Both analytical and preparative injection valves are available to fit different sample volumes and flow rates
  • Various rotor seal materials are available, including Vespel, Tefzel, and PEEK, to accommodate a range of sample pH
  • Choice of stainless steel or PEEK flowpath
  • The right mix of factory-cut and finished sample loops of highest quality for your application needs
  • Color-coded manual syringes with a wide selection of volumes, improved scale readability, and optimal accuracy and
More Product Information