CrossLab Inert Torches and Injectors for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3285-80034 G4912-80012 G4912-80013 G3285-80035 G3285-80085 G4912-80010 G4912-80011
CAAGG328580034KT 6205.25 CAD
CAAGG328580034 CAAGG491280012 CAAGG491280013 CAAGG328580035 CAAGG328580037 CAAGG328580085 CAAGG491280010 CAAGG491280011
CrossLab Inert Torches and Injectors for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Demountable torch, PFA injector holder and inert sapphire and platinum injectors for inert ICP-MS kit.
  • PFA injector holder uses a push fit, O-ring free seal to provide a secure connection to the base of the demountable quartz torch, and ensures easy removal when required
  • Sapphire injectors are commonly recommended for organic solvents
  • Platinum injectors recommended for HF digests, aggressive acids and ultratrace analysis
  • Injectors available in 1.5 mm Int.Ø recommended for use with organic solvents and 2.5 mm recommended for aggressive acids
  • PFA injector holder manufactured from the highest purity grade PFA resin for the lowest blank contribution, ensuring it is suitable for semiconductor applications.
The inert PFA sample introduction kit for Agilent ICP-MS instruments includes a PFA injector holder, fitted to a semi-demountable quartz torch. The semi demountable quartz torch fits onto the injector holder using a push fit, O-ring free seal to provide a secure seal that can be easily removed, without any risk of the O-ring being set in place.
Injectors are available in either sapphire or platinum, and in both 1.5 and 2.5 mm Int.Ø.