CrossLab Headspace Sampler Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
1535-4952 19258-20830 19258-20870 2321700003 2322590004 2322590005 2322700011 301-013-HSP 301-016-HSP 301-220-HSP 301-015-HSP 0100-2595 0100-2594 G4556-63825 4177-0607 G4556-80106 G1888-60702 G3520-20210 G4556-60019 G4556-67010 G4556-67011 G4556-80105 G4556-80108 G4556-80126 5183-4801 G1888-60703 G4556-67012 2321700004 5190-2266
CA1535-4952EA 284.25 CAD
CA1535-4952 CA19258-20830 CA19258-20870 CA2321700003 CA2322590004 CA2322590005 CA2322700011 CA301-013-HSP CA301-016-HSP CA301-220-HSP CA301-015-HSP CA0100-2595 CA0100-2594 CAG4556-63825 CA4177-0607 CAG4556-80106 CAAGG188860702 CAAGG352020210 CAAGG455660019 CAAGG455667010 CAAGG455667011 CAAGG455680105 CAAGG455680108 CAAGG455680126 CAAG5183-4801 CAAGG188860703 CAAGG455667012 CAAGG455690500 AG2321700004 AG5190-2266
CrossLab Headspace Sampler Supplies for GC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Find the headspace supplies you need for your Agilent 7697 or G1888 headspace sampler.
  • Built-in pneumatics for superior control and easier setup;Proven valve and loop sampling technology
  • Fully-automatic sample vial leak checking and available bar-code reader
  • Instrument control software that is fully integrated in Agilent data systems
  • Programmable resource-conserving instrument scheduler
Headspace sampling allows the introduction of volatile compounds to the GC or GC/MS from virtually any sample matrix while leaving unwanted components in a disposableMore Product Information