CrossLab Graphite Furnace AAS Tubes and Platforms, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
6310001200 6310003700 6310001100 6310001300
CA6310001200PK 905.39 CAD
CA6310001200 CAAG6310003700 CAAG6310001100 CAAG6310001300
CrossLab Graphite Furnace AAS Tubes and Platforms, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Graphite tubes are tested to ensure each batch meets demanding performance specifications.
  • High-purity graphite reduces spurious absorption from trace contaminants and improves signal-to-noise
  • Every tube is individually checked for reproducible and reliable results
  • Perform fewer repeats and increase productivity
Agilent graphite tubes are batch tested to ensure each batch passes they exceed our demanding performance requirements specifications for contamination, sensitivity, precision, electrical resistance, and lifetime. Partitioned tubes provide are suited for general purpose work and the best detection limits with samples that have with low background. They are also ideal for organic solvents solutions or samples with low viscosity samples that may spread and degrade precision. Omega platform tubes combine ease-of-use with perfect atomization The integrated platform ensures the perfect thermal profile for good signal-to-noise ratios and best detection limits. The solid pyrolytic platforms and plateau tubes provide continuity with older methods.