CrossLab Gas Tubing for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3270-65035 G3280-67022 G3280-67023 G3138-65138 G3280-20663 G3280-60160 G3280-60163 G3666-67026
CAG3270-65035EA 432.19 CAD
CAG3270-65035 CAAGG328067022 CAAGG328067023 CAAGG313865138 CAAGG328020663 CAAGG328060160 CAAGG328060163 CAAGG366667026
CrossLab Gas Tubing for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Tubing for the nebulizer (or carrier), make-up, dilution or cell gas supplies, sleeves and connectors.
  • PTFE tubing is used to introduce the carrier, make-up, dilution or cell gases to the sample introduction system or the ORS reaction cell
  • Replacement PTFE tubing, sleeves and connectors are available so you can ensure a gas tight and leak free supply of gases for your application
  • Low out-gassing plastic tubing for Agilent ICP-MS gas line ensures low backgrounds and minimizes any potential for contamination
PTFE tubing is used to introduce the carrier,More Product Information