CrossLab Gas Cylinder Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0100-0111 0100-0118 0100-0119 0460-1266 5180-4143 5183-1941 5183-4641 5183-4642 5183-4645 5183-4644 0100-0110 0101-1535 0101-1536 19246-60560 19246-60570 19246-60690 5180-4145 5183-4643 5188-5374 G4218-60100 0101-1400
CA0100-0111EA 88.58 CAD
CA0100-0111 CA0100-0118 CA0100-0119 CA0460-1266 CA5180-4143 CA5183-1941 CA5183-4641 CA5183-4642 CA5183-4645 CA76514-896 CAAG0100-0110 CAAG0101-1535 CAAG0101-1536 CAAG1924660560 CAAG1924660570 CAAG1924660690 CAAG5180-4145 CAAG5183-4643 CAAG5188-5374 CAAGG421860100 AG0101-1400
CrossLab Gas Cylinder Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent gas cylinder supplies include economical brass regulators, wall brackets, and NPT adapters.
  • Dual-stage regulators provide more precise and consistent pressure control than single-stage regulators;Maximum inlet pressure: 3000 psig
  • Operating temperature range: ‒40 to 165 °F
  • Swagelok regulator for female flare NPT adapter available in ¹/₈ or ¹/₄ brass connector
Agilent gas cylinder supplies include regulators that provide proper gas pressure control for gas chromatography. These brass-body regulators are available for US only and follow the Compressed Gas Association (CGA) connection guidelines. Each gas regulator comes with ¹/₈ inch pipe thread fitting.