CrossLab Fraction Collector Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5022-6532 5022-6538 5022-6539 G1364-68711 G1364-68712 G1364-87202 G1367-87200 5022-6533 5022-6534 5042-6459 5190-9090 5190-9091 5042-6458 5042-6470 5190-9092 5190-9093 G1364-60111 G1364-44610 G1364-44612 G1364-44613 G1364-87201 G9321-60015 G9321-60025 G9321-60055 G9321-60129 G2255-68700 5023-2872 5023-2874 5023-2875 5023-2878 5023-2882 5023-2883 5043-1826 5067-6840 5067-6857 5188-8053 G1364-60621 G1364-68601 G1364-68602 G1364-68603 G1364-68604 G1364-68605 G1364-68723 G1364-84522 G1364-84536 G1364-84543 G1364-84544 G1364-84545 G1364-86711 G1364-87304 G1364-87305 G1364-87306 G1972-60025 G9321-60035 G9321-60045 G9321-60058 G9321-60085 G9321-60131 G9321-60951 G9321-60952 G9321-60954
CA5022-6532PK 74.46 CAD
CA5022-6532 CA5022-6538 CA5022-6539 CAG1364-68711 CAG1364-68712 CAG1364-87202 CAG1367-87200 CA76514-628 CA76514-630 CA76514-634 CA5190-9090 CA5190-9091 CA5042-6458 CA5042-6470 CA5190-9092 CA5190-9093 CAG1364-60111 CAAGG136444610 CAAGG136444612 CAAGG136444613 CAAGG136487201 CAAGG932160015 CAAGG932160025 CAAGG932160055 CAAGG932160129 CAG2255-68700 CAAG5023-2872 CAAG5023-2874 CAAG5023-2875 CAAG5023-2878 CAAG5023-2882 CAAG5023-2883 CAAG5043-1826 CAAG5067-6840 CAAG5067-6857 CAAG5188-8053 CAAGG136460621 CAAGG136468601 CAAGG136468602 CAAGG136468603 CAAGG136468604 CAAGG136468605 CAAGG136468723 CAAGG136484522 CAAGG136484536 CAAGG136484543 CAAGG136484544 CAAGG136484545 CAAGG136486711 CAAGG136487304 CAAGG136487305 CAAGG136487306 CAAGG197260025 CAAGG932160035 CAAGG932160045 CAAGG932160058 CAAGG932160085 CAAGG932160131 CAAGG932160951 CAAGG932160952 CAAGG932160954
CrossLab Fraction Collector Supplies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A variety of supplies for a range of fraction collectors to suit your purification needs.
  • All supplies manufactured with highest precision to ensure a perfect fit
  • Various dimensions of well plates, collection tubes and trays to suit a wide variety of applications and sample volumes are available
  • ESD-striped tubing helps avoid accumulation electrostatic charges
Agilent provides a variety of parts and supplies for fraction collectors, including test tubes, tube containers and drawers, well plates and trays, and tubing and tubing kits, as well as delay calibrant, ensuring enhanced performance and capability of your fraction collector.
Agilent supplies for fraction collectors are part of a comprehensive portfolio of flexible and reliable solutions for sample purification by LC. Various fraction collectors are available with maximum flow rates from 4 µl/min to 200 ml/min.