CrossLab Demountable Torch Supplies for 700 Series, Vista Series and Liberty ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
2010070790 2010072900 9910084700 9910107100 2010083400 7910044500 9910056800
CAAG2010070790EA 212.25 CAD
CAAG2010070790 CAAG2010072900 CAAG9910084700 CAAG9910107100 CAAG2010083400 CAAG7910044500 CAAG9910056800 CAAG9910106300
CrossLab Demountable Torch Supplies for 700 Series, Vista Series and Liberty ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Injectors, quartz tubes, O-rings, and other supplies for demountable torches.
  • Demountable torch for 700 series, Liberty, and Vista ICP-OES light easily and stay lit, reducing downtime
  • A wide variety of injector sizes and materials available to meet application needs
  • Semi and fully demountable torches allow replacement of individal parts leading to cost savings for the lab
Demountable torches provide ease of maintenance and reduced costs due to having the flexibility to replace specific components, instead of a complete torch. Demountable torchMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable