CrossLab Column Thermostat Accessories for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5062-8588 G1316-87303 5063-6526 G7116-68003 G7116-68004 G7116-60013 5001-3702 5043-1465 5067-5917 5067-5963 5067-6654 5067-6849 G1316-60001 G7163-60002
CA5062-8588PK 273.55 CAD
CA5062-8588 CA76515-212 CA5063-6526 CAG7116-68003 CAG7116-68004 CAAGG711660013 CAAG5001-3702 CAAG5043-1465 CAAG5067-5917 CAAG5067-5963 CAAG5067-6654 CAAG5067-6849 CAAGG131660001 CAAGG716360002
CrossLab Column Thermostat Accessories for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Supplies to ensure the performance of the thermostatted column compartment in HPLC systems.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Column Int.Ø tags allow you to track various column properties and usage parameter including but not limited to column identity, lot and batch number, the last injection date, number of injections, and the max temperature used
  • Column Int.Ø tags ensure traceability of analysis, documenting columns and conditions for routine analysis easy and adding confidence to any analysis
Agilent provides a comprehensive range of high-quality column thermostat accessories and supplies, including column tags and heat exchangers, to support your efforts in increasing your analytical results.
All parts are manufactured to Agilent's highest levels of quality and have been tested in Agilent instruments to ensure they work perfectly with the rest of the system.