CrossLab Alignment Interfaces, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G7100-60210 G7100-60230 G1600-60210 G1600-60230 G1600-60310 G1600-60330 G7100-60310 G7100-60330 G7100-60400
CAAGG710060210EA 665.1 CAD
CAAGG710060210 CAAGG710060230 CAAGG160060210 CAAGG160060230 CAAGG160060310 CAAGG160060330 CAAGG710060310 CAAGG710060330 CAAGG710060400
CrossLab Alignment Interfaces, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Together with the capillary cassette, alignment interfaces simplify capillary exchange.
  • The quick-change cassette allows capillary exchange in less than a minute
  • The cassette and interfaces accept all commercially available capillaries with a 365 µm Ext. Ø
Agilent alignment interfaces, with the capillary cassette, simplify capillary exchange, protect the fragile detection window, and ensure exact alignment of the window in the detector. Alignment interfaces are an integral part of the Agilent diode-array detection (DAD) system. These interfaces contain optical slits that are precisely matched to the capillary inner diameter for optimized sensitivity and linear detection range.