VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR
VWR®
46610-740A 66009-850A 66010-539A 89239-008A 82028-410A 66009-864A 46610-742A 82028-408A
46610-742ACS 770.96 CAD
46610-742A 89239-008A 66010-539A 66009-864A 82028-408A 66009-850A 46610-740A 82028-410A
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Vials
11 mm crimp top, wide neck opening design, allows easy filling, requires inserts with a diameter of 6 mm.
  • Aluminum crimp closures provide a secure leak-resistant seal
  • Aluminum seals must be applied with a crimping tool
  • Spring promotes alignment and ensures insert seals against septa to help guard against evaporation loss
Wide opening vials feature 40% larger neck opening than standard opening vials, for easier access and reduced needle damage.