Specimen Containers with Caps, Caplugs
Supplier: Caplugs
240-3007-060 240-3010-060 240-5601-G80 240-5603-G80
76480-220CT 285.63 CAD
76480-220 76480-226 76499-438 76499-442
Specimen Containers with Caps, Caplugs
Sample Containers General Purpose Containers
Free standing containers that are ideal for collecting/transporting general purpose specimens. The containers are constructed of high-clarity polypropylene copolymer with high density polyethylene screw caps.
  • All containers are sold with caps
  • All containers are available with patient information labels
  • All containers are natural color
Designed for storing biological fluids and pathology specimens.