Allegra V-15R Benchtop Centrifuges, Beckman Coulter
Supplier: Beckman Coulter
C63124 C63125 C63375 C63380 C63381 C63432 C63433 C63444 C63445 C63450 C69866 C69867
CA76521-700EA 16102.94 CAD
CA76521-700 CA76521-702 CA76521-704 CA76521-706 CA76521-708 CA76521-722 CA76521-724 CA76521-730 CA76521-732 CA76521-734 76628-204 76628-206
Allegra V-15R Benchtop Centrifuges, Beckman Coulter
Centrifuges Benchtop Centrifuges
The Allegra V-15R benchtop centrifuges bring more versatility to the laboratory bench. These compact, refrigerated centrifuges are ideal for evolving laboratories.
  • Free up precious benchtop space
  • Support multiple workflows with just one instrument
  • Advanced temperature management
  • Wide choice of centrifuge packages available to suit various applications
The Allegra V-15R centrifuges maintain a consistent temperature, allowing you to spin faster and longer without overheating your samples. All while Rapid Temp and Eco Mode allow for even more efficiency and functionality.
The Allegra V-15R centrifuges are designed to be a workhorse in the laboratory. Operation is simple and seamless thanks to an easy-to-use interface and ergonomic design. With 10 rotor options at your fingertips, you will be ready and able to support multiple workflows with just one instrument.
10 rotors and a comprehensive selection of adapters allows for a variety of applications, from cell and blood separation to high throughput screening. Its compact footprint frees up space on your bench and its ≤55 dBA (at fullspeed) makes it a quiet companion in the lab.
Operation is simple and seamless with soft-touch lid closure, 50 programmable runs and an easy-to-use illuminated interface.