SterilGARD Biosafety Containment Cabinet, for CytoFLEX SRT, Beckman Coulter
Supplier: Beckman Coulter
C71892 C71892
CA76521-328EA 111557.56 CAD
76521-328 CA76521-328
SterilGARD Biosafety Containment Cabinet, for CytoFLEX SRT, Beckman Coulter
Fume Hoods and Enclosures Biological Safety Cabinets
Class II Type biosafety containment cabinet designed specifically for the CytoFLEX SRT in collaboration with Baker Corporation, including aerosol evacuation functionality.
  • Personnel protection or containment by an intake air velocity of no less than 100 feet per minute (fpm) through the front access opening
  • Product protection by the HEPA (high efficiency particulate air) filtered downflow air inside the cabinet work area
  • Environmental protection by allowing only HEPA filtered to be exhausted to the room
For indoor use only.
Certifications: CE, RUO, EN 12469, BS EN 12469, SNS 12469, NF-095, YY-0569, JIS K 3800, AS 1807.1:2009.