CytoFLEX SRT Bench Top Cell Sorter, Beckman Coulter

Supplier: Beckman Coulter
C71883 C71884 C71885 C71886 C71887 C71888 C71889
CA76521-314EA 697231.38 CAD
CA76521-314 CA76521-316 CA76521-318 CA76521-320 CA76521-322 CA76521-324 CA76521-326
CytoFLEX SRT Bench Top Cell Sorter, Beckman Coulter
Cell Counters and Flow Cytometers Flow Cytometers
If you need an approachable instrument for sorting multiple populations to generate high quality material for downstream assays, the CytoFLEX SRT benchtop cell sorter is quick to learn and easy to operate. This extension of the CytoFLEX platform is built on the same optical features as the flow cytometer, but equally important, it continues to adhere to the principles that made multicolor applications accessible to many biomedical research, core research, pharma and biotechs.

  • Exquisite sensitivity for multicolor applications, up to 15 fluorescent colors to define populations
  • Parameter matched to the CytoFLEX S V-B-Y-R series
  • Intuitive software to facilitate multicolor analysis, CytExpert for CytoFLEX SRT
  • Uses technology to increase ease of use by automating sort stream setup and maintenance throughout the sort
  • 4-way sorting, mixed mode sorting, ability to catch aborts and preserve precious cells
  • High recovery and post sort viability
  • Accurate single-cell deposition using the Cyclone Movement System, including index sorting
  • Sort into tubes, slides, or microplates

CytoFLEX SRT cell sorter is a benchtop sorter. It is capable of meeting requirements for a wide range of sorting needs. And like the CytoFLEX Platform, it includes innovative technologies that simplify the setup and operation, empowering investigators to focus on the research questions.

The Violet-Blue-Yellow Green-Red (V-B-Y-R) series has 15 fluorescent detectors when fully activated. The fully activated instrument includes five fluorescent channels from the 405 nm (Violet) laser, two from the 488 nm (Blue) laser, five from the 561 nm (Yellow Green) laser, and three from the 638 nm (Red) laser. It can be purchased with as few as five with an option to activate additional lasers and detectors with an activation key.

It is capable of complex sort logic with different combinations of sort settings on each of four streams, including the ability to catch aborts of one of the other streams. The CytoFLEX SRT cell sorter is designed for reliability and supported by teams of experts, remote support is ready using BeckmanConnect for on demand access to our technical specialists, perform many routine maintenance procedures independently.

Additional specifications:
Nozzle diameter: 100 µm
Signal processing: Digital system with 7-decade display
Drop dive frequency: Automatically set, manually adjustable between 30000 - 35000 Hz
Sorting performance: With a target population of 5% and a threshold rate of <10,000 events per second, >99% purity and >80% of Poisson's expected yield
Sorting modes: 4-way sorting; enrichment or purity or single, ability to catch aborts
Collection vessels: 12×75 mm tubes, 15 ml conical tubes, 96-well plates, 96-deep-well plates, 384-well plates, slides
Optical filter: 450/45, 525/40 (2), 585/42, 610/20 (2), 660/10 (2), 675/30, 690/50, 710/50, 712/25, 780/60 (3)

Certifications: CE, RUO.

Accessories information: 76521-332: This bundle pack includes consumables required to operate the CytoFLEX SRT Cell Sorter.
76521-334: This bundle is designed for 1-year of preventive maintenance following the routine maintenance schedule. See CytoFLEX SRT Cell Sorter Instructions for Use for detail.

Ordering information: CytoFLEX SRT is supplied with CyExpert for SRT, computer, monitor and keyboard. An optional Biosafety Cabinet including aerosol evacuation or a standalone Aerosol Evacuation Unit is available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR