Bond Elut Plexa Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
A3969010 A3969030B A4969010 A4969030HF
CAAGA3969010EA 715.57 CAD
CAAGA3969010 CAAGA3969030B CAAGA4969010 CAAGA4969030HF
Bond Elut Plexa Plates, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut Plexa offers simple extraction, minimal method development, and improved cleanliness.
  • Nonpolar retention mechanism
  • Improved extract cleanliness minimizes sample matrix interferences
  • Simple methods are amenable to a very broad range of analytes
  • Fast flow, reproducible performance, and ease-of-use
Bond Elut Plexa offers easy-to-use methods for general purpose extraction. A unique polymeric architecture with a nonretentive, hydroxylated, amide-free surface, and a nonpolar PS-DVB core retains small molecules. Binding of proteins and lipids on the polymer surface is minimized, giving cleaner samples and reduced ion suppression.
Bond Elut Plexa offers easy-to-use methods for general purpose extraction. A unique polymeric architecture with a nonretentive, hydroxylated, amide-free surface, and a nonpolar PS-DVB core retains small molecules. Binding of proteins and lipids on the polymer surface is minimized, giving cleaner samples and reduced ion suppression.
Plexa is ideal for high-throughput assays requiring validated performance with minimal method development. The standard nonpolar retention mechanism is applicable to almost any analyte type. Plexa Polymeric SPE is also available in ion exchange SPE formats, Plexa PCX SPE for cation exchange, and Plexa PAX for anion exchange.